Skip to Content
Merck
All Photos(3)

Key Documents

114421

Sigma-Aldrich

Bromothymol Blue sodium salt

ACS reagent

Synonym(s):

3′,3′′-dibromothymolsulfonephthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27Br2NaO5S
CAS Number:
Molecular Weight:
646.36
Beilstein:
6887926
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

crystalline powder
powder or crystals

technique(s)

titration: suitable

visual transition interval

6.0-7.6, yellow to blue

solubility

water: 0.1%, clear to very slightly hazy

λmax

392 nm
615 nm

ε (extinction coefficient)

≥35000 at 614-618 nm in 0.1 M NaOH at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

SMILES string

CC1=C(C=C(C(=C1Br)O)C(C)C)C(=C2C=C(C(=O)C(=C2C)Br)C(C)C)C3=CC=CC=C3S(=O)(=O)[O-].[Na+]
[Na+].CC(C)c1cc(c(C)c(Br)c1O)C(=C2\C=C(C(C)C)C(=O)C(Br)=C2C)\c3ccccc3S([O-])(=O)=O

InChI

InChI=1S/C27H28Br2O5S.Na/c1-13(2)18-11-20(15(5)24(28)26(18)30)23(17-9-7-8-10-22(17)35(32,33)34)21-12-19(14(3)4)27(31)25(29)16(21)6;/h7-14,30H,1-6H3,(H,32,33,34);/q;+1/p-1/b23-21+;
1S/C27H28Br2O5S.Na/c1-13(2)18-11-20(15(5)24(28)26(18)30)23(17-9-7-8-10-22(17)35(32,33)34)21-12-19(14(3)4)27(31)25(29)16(21)6;/h7-14,30H,1-6H3,(H,32,33,34);/q;+1/p-1/b23-21+;

InChI key

NMKFVGALBGZKGW-FKWCIMQXSA-M
NMKFVGALBGZKGW-FKWCIMQXSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Bromothymol Blue has been used for freshwater pH measurements over a broad range of buffer intensities.

Biochem/physiol Actions

Bromothymol Blue is a pH-indicator. It is also used for carbonic anhydrase assay due to the conversion of carbon dioxide to bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrophilic region of lecithin membranes studied by bromothymol blue and effects of an inhalation anesthetic, enflurane.
Mashimo T, et al.
Proceedings of the National Academy of Sciences of the USA, 76, 5114-5118 (1979)
Biochemical properties of a new a-carbonic anhydrase from the human pathogenic bacterium, Vibrio cholerae.
Del Prete S, et al.
Journal of Enzyme Inhibition and Medicinal Chemistry, 29, 23-27 (2014)
Evaluation of indicator-based pH measurements for freshwater over a wide range of buffer intensities.
Yuan S and DeGrandpre MD
Environmental Science & Technology, 42, 6092-6099 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service