Skip to Content
Merck
All Photos(1)

Key Documents

MR-051

Sigma-Aldrich

M2 medium

EmbryoMax M2 Medium (1X), Liquid, with Phenol Red & Hyaluronidase

Synonym(s):

Cell Culture Medium, Growth Medium M2, M2 Medium Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | embryo: suitable
cell culture | stem cell: suitable

input

sample type: mouse embryo(s)

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service