Skip to Content
Merck
All Photos(1)

Key Documents

MABS197

Sigma-Aldrich

Anti-PTP1B Antibody, clone FG6-1G

clone FG6-1G, from mouse

Synonym(s):

Tyrosine-protein phosphatase non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
FG6-1G, monoclonal
application:
IP
WB
species reactivity:
human
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:
10

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FG6-1G, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPN1(5770)

General description

PTP1B is the prototypic member of the protein tyrosine phosphatase (PTP) family, which comprises at least 37 proteins. The family is characterized by a catalytic phosphatase domain of approximately 240 amino acids, and includes both transmembrane and cytosolic enzymes. The PTPs have high substrate specificity for phosphotyrosyl proteins. At the primary sequence level, PTPs share little similarity with the protein serine phosphatases, protein threonine phosphatases, or the acid and alkaline phosphatases. PTP1B is a negative regulator of insulin and leptin signal transduction and may be a potential therapeutic target for diabetes and obesity. Insulin also influences the expression of splice variants of PTP1B.

Specificity

This antibody recognizes the catalytic domain of PTP1B.

Immunogen

Epitope: Catalytic domain
Recombinant protein corresponding to the catalytic domain of human PTP1B.

Application

Anti-PTP1B Antibody, clone FG6-1G is an antibody against PTP1B for use in Western Blotting, Immunoprecipitation.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
Western Blot Analysis: A representative lot was used to detect PTP1B in Jurkat cell lysate.

Immunoprecipitation Analysis: A representative lot from an independent laboratory detected PTP1B in A431 cell lysate (Gulati, P. et al. (2004). EMBO Rep. 5(8):812-817.).

Quality

Evaluated by Western Blot in TF-1 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected PTP1B in 10 µg of TF-1 cell lysate.

Target description

~50 kDa observed

Linkage

Replaces: 04-1140

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
TF-1 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giampaolo Minetti et al.
The Biochemical journal, 377(Pt 2), 489-497 (2003-10-07)
One of the most intensively studied post-translational modifications of erythrocyte proteins is the phosphorylation of tyrosine residues of band 3, which is strictly regulated in vivo by PTKs (protein-tyrosine kinases) and PTPs (protein-phosphotyrosine phosphatases). Two PTKs (p72(syk) and p56/53(lyn)) and
Jennifer J Schwarz et al.
Life science alliance, 4(11) (2021-09-17)
B cell antigen receptor (BCR) signaling is initiated by protein kinases and limited by counteracting phosphatases that currently are less well studied in their regulation of BCR signaling. Here, we used the B cell line Ramos to identify and quantify
Malena Zahn et al.
Carcinogenesis, 42(4), 517-527 (2021-01-01)
The neoplastic Hodgkin/Reed-Sternberg (HRS) cells of classical Hodgkin lymphoma (cHL) depend on chronic activation of the Janus kinase (JAK)/signal transducer and activator of transcription (STAT) signalling pathways to maintain survival and proliferation. Accumulating reports highlight the importance of the inactivation
Fawaz G Haj et al.
The Journal of biological chemistry, 280(15), 15038-15046 (2005-02-09)
Protein-tyrosine phosphatase 1B (PTP1B) is an important negative regulator of insulin and leptin signaling in vivo. Mice lacking PTP1B (PTP1B-/- mice) are hyper-responsive to insulin and leptin and resistant to diet-induced obesity. The tissue(s) that mediate these effects of global
Fawaz G Haj et al.
The Journal of biological chemistry, 278(2), 739-744 (2002-11-09)
Receptor tyrosine kinases (RTKs) are key regulators of cellular homeostasis. Based on in vitro and ex vivo studies, protein tyrosine phosphatase-1B (PTP1B) was implicated in the regulation of several RTKs, yet mice lacking PTP1B show defects mainly in insulin and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service