Skip to Content
Merck
All Photos(3)

Key Documents

MABF3054

Sigma-Aldrich

Anti-SARS CoV-2 RBD mutant N501Y Antibody, clone 8A6

Synonym(s):

Spike glycoprotein Mutant N501Y

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
8A6, monoclonal
application:
ELISA
WB
species reactivity:
vaccinia virus
technique(s):
ELISA: suitable
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8A6, monoclonal

mol wt

calculated mol wt 141 kDa

purified by

using protein G

species reactivity

vaccinia virus

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

vaccinia virus ... S(43740568)

Specificity

Clone 8A6 is a mouse monoclonal antibody that detects N501Y mutant form of of SARS-CoV-2 spike protein.

Immunogen

KLH-conjugated linear peptide segment surrounding N501Y mutant site.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant SARS-CoV-2 Spike RBD (N501Y) protein .

Western Blotting Analysis: A 1:500 dilution of this antibody detected recombinant SARS-CoV-2 RBD mutant N501Y protein.

Tested Applications

ELISA Analysis: Serial dilutions of this antibody detected various recombinant SARS-CoV-2 protein samples

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Spike glycoprotein (UniProt: P0DTC2; also known as S glycoprotein, E2, Peplomer protein) is encoded by the S (also known as 2) gene (Gene ID: 43740568) in Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). The SARS-CoV-2 is a positive-strand RNA virus that causes severe respiratory syndrome in human. The mature SARS-CoV-2 contains 4 structural proteins: Envelope (E), Membrane (M), Nucleocapsid (N), and the Spike protein (S). E and M proteins help in viral assembly and N protein is needed for RNA synthesis. The S protein is a single-pass type I, homotrimeric, membrane glycoprotein that is responsible for virus binding and entry into host cell. It is synthesized with a signal peptide (aa 1-12), which is subsequently cleaved off to generate the mature protein that contains an extracellular domain (aa 13-1213), a transmembrane domain (aa 1214-1234), and a cytoplasmic domain 1235-1273). The S protein is further processed into S1 (aa 13-685) and S2 (aa 686-1273) subunits by host cell furin. The presence of a furin polybasic cleavage site in S protein from SARS-CoV-2 sets it apart from S protein in SARS-CoV that possesses a monobasic S1/S2 cleavage site. The S1 subunit has the receptor binding domain (RBD; aa 319-541) that mediates entry of SARS-CoV-2 into sensitive cells through the peptidase domain of host Angiotensin-converting enzyme 2 (ACE2) with high affinity (KD = 15 nM). The S2 domain (aa 686-1213), which is reported to be well conserved, is responsible for membrane fusion. Over the course of the pandemic, SARS-CoV2 has consistently mutated giving rise to various mutants. Clone 8A6 recognizes SARS-CoV-2 spike protein with N501Y mutation. Several variants, including B.2.2.7, B.1.351, and P.1 share this mutation on the receptor binding domain (RBD) and is a more infectious variant. RBD with N501Y mutation exhibits a stronger interaction with ACE2 receptor and slower dissociation rate. (Ref.: Liu, Y., et al. (2022). Nature. 602(7696); 294-299; Tian, F., et al. (2021). eLife. 10; e69091).

Physical form

Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service