Skip to Content
Merck
All Photos(1)

Key Documents

MAB8183

Sigma-Aldrich

Anti-EBV MA-gp350/250 Antibody, clone 2L10

clone 2L10, Chemicon®, from mouse

Synonym(s):

Membrane Antigen gp350/250

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2L10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

Specificity

MA - gp 350/250

Application

Detect EBV MA-gp350/250 using this Anti-EBV MA-gp350/250 Antibody, clone 2L10 validated for use in ELISA, IF & WB.
Immunofluorescence

ELISA (Non neutralizing)

Immunoblotting (weak)

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M phosphate buffer, pH 7.6, 0.25M NaCl, with 0.1% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen Vallhov et al.
Journal of immunology (Baltimore, Md. : 1950), 186(1), 73-82 (2010-11-26)
Exosomes are nano-sized membrane vesicles released from a wide variety of cells, formed in endosomes by inward budding of the endosomal limiting membrane. They have immune stimulatory-, inhibitory-, or tolerance-inducing effects, depending on their cellular origin, which is why they
Cellular microRNAs 200b and 429 regulate the Epstein-Barr virus switch between latency and lytic replication.
Ellis-Connell, AL; Iempridee, T; Xu, I; Mertz, JE
Journal of virology null
Febri Gunawan Sugiokto et al.
mBio, 15(7), e0079524-e0079524 (2024-06-14)
Epstein-Barr virus (EBV) is a ubiquitous human tumor virus that establishes lifelong, persistent infections in B cells. The presence of EBV in cancer cells presents an opportunity to target these cells by reactivating the virus from latency. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service