Skip to Content
Merck
All Photos(1)

Documents

MAB1378

Sigma-Aldrich

Anti-Integrin α6 Antibody, clone NKI-GoH3

clone NKI-GoH3, Chemicon®, from rat

Synonym(s):

CD49f

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

NKI-GoH3, monoclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA6(3655)

Related Categories

General description

Integrins are a family of dimeric, transmembrane proteins that mediate cell-cell and extracellular matrix adhesion. Signals transduced by integrins play a role in many biological processes, including cell growth, differentiation, migration and apoptosis. The integrin family is composed of at least 15 alpha and 8 beta subunits that may form over twenty different alpha-beta non-covalently bound dimeric combinations on the cell surface. The alpha subunits all have some homology to each other, as do the beta subunits. Both of the subunits contribute to the binding of the ligand. Integrin alpha subunits contain seven weak sequence repeats in the N-terminal region, which may be important in ligand binding, and have been predicted to fold cooperatively into a single beta-propeller domain with seven beta-sheets. In normal tissue, Integrin alpha-6/beta-1 is a receptor for laminin on human platelets. Integrin alpha-6/beta-4 is a receptor for laminin in epithelial cells and it plays a critical structural role in the hemidesmosome. Various disease states involving epithelial cells have been shown to be associated with alterations in alpha 6 integrin-containing heterodimers.

Specificity

Directed against the Cdw49f-antigen, 120kDa, (GP Ic or VLA-6 alpha-chain), which can form distinct complexes with either the CD29-antigen (GP 11a or VLA beta-chain), resulting in the VLA-6 (alpha-6 beta-1) complex, which is expressed on human platelets, or with the beta-4 integrin resulting in the alpha-6 beta-4 complex which is expressed on various human epithelial cells. The monoclonal antibody reacts with platelets, megakaryocytes, T lymphocytes and common Acute Lymphoblastic Leukemia cells (alpha-6 beta-1). In immunohistology the monoclonal antibody reacts with epithelial cells of a variety of tissues, peripheral nerves, microvascular endothelial cells, placenta cyto- and syncytotrophoblasts. Platelet VL-6 functions as a laminin receptor and platelet adhesion to laminin is inhibited by GoH3.

Immunogen

Mammary tumor cells from BALB/c mice.

Application

Detect Integrin α6 using this Anti-Integrin α6 Antibody, clone NKI-GoH3 validated for use in FC, IH.
Flow cytometry: 1:10

Immunohistology on frozen tissue sections: 1:25-1:50

Immunoprecipitation (Sonnenberg, et al. 1990)

Inhibition of laminin binding:40μg/ml.



Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Human intestine tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaelyn D Sumigray et al.
Developmental cell, 45(2), 183-197 (2018-04-25)
The adult mammalian intestine is composed of two connected structures, the absorptive villi and the crypts, which house progenitor cells. Mouse crypts develop postnatally and are the architectural unit of the stem cell niche, yet the pathways that drive their
Integrin regulation by vascular endothelial growth factor in human brain microvascular endothelial cells: role of alpha6beta1 integrin in angiogenesis.
Lee, TH; Seng, S; Li, H; Kennel, SJ; Avraham, HK; Avraham, S
The Journal of Biological Chemistry null
Michael E Todhunter et al.
Nature methods, 12(10), 975-981 (2015-09-01)
Reconstituting tissues from their cellular building blocks facilitates the modeling of morphogenesis, homeostasis and disease in vitro. Here we describe DNA-programmed assembly of cells (DPAC), a method to reconstitute the multicellular organization of organoid-like tissues having programmed size, shape, composition
Yuki Fujii et al.
Journal of cell science, 129(7), 1512-1522 (2016-02-14)
Placement of a tag sequence is usually limited to either terminal end of the target protein, reducing the potential of epitope tags for various labeling applications. The PA tag is a dodecapeptide (GVAMPGAEDDVV) that is recognized by a high-affinity antibody
Somatic correction of junctional epidermolysis bullosa by a highly recombinogenic AAV variant.
Melo, SP; Lisowski, L; Bashkirova, E; Zhen, HH; Chu, K; Keene, DR; Marinkovich, MP; Kay, MA; Oro, AE
Molecular Therapy null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service