Skip to Content
Merck
All Photos(1)

Key Documents

ABN462

Sigma-Aldrich

Anti-Dopamine D2 Receptor Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Dopamine D2 Receptor, D2R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
species reactivity:
mouse
technique(s):
immunohistochemistry: suitable
citations:
5
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Drd2(13489)

General description

The Dopamine D2 Receptor is one of the five types (D1 to D5) of receptors for dopamine. The activity of this receptor is mediated by G proteins which inhibit adenylyl cyclase. 2 isoforms are reported for the D2 Dopamine Receptor, Isoform 1 and isoform 2 which are expressed in the pituitary and brain; Isoform 1 is expressed seven times more than isoform 2 in the caudate nucleus.

Specificity

This antibody binds to the cytoplasmic third intracellular loop.

Immunogen

Epitope: cytoplasmic third intracellular loop
Recombinant protein corresponding to the cytoplasmic domain of mouse Dopamine D2 Receptor.

Application

Anti-Dopamine D2 Receptor Antibody is an antibody against Dopamine D2 Receptor for use in Immunohistochemistry.
Immunohistochemistry Analysis: A representative lot detected Dopamine D2 Receptor in WT mouse striatum tissue (Trifilieff, P., et al. (2013). Molecular Psychiatry. 18:102-1033).
Immunohistochemistry Analysis: A representative lot detected Dopamine D2 Receptor in wild-type and not in D2R knockout mouse sriatum tissue (Trifilieff, P., et al. (2011). BioTechniques. 50:111-118).
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Immunohistochemistry in mouse striatum tissue.

Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected Dopamine D2 Receptor in mouse striatum tissue.

Target description

50.9 kDa calculated

Physical form

Affinity
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérôme Jeanblanc et al.
Brain : a journal of neurology (2024-05-05)
The use of psilocybin to treat alcohol use disorder is very promising, but the mechanisms of action remain poorly understood. We combined behavioral, pharmacological and gene expression analyses to decipher the mechanisms of action of psilocybin, for the first time
Ying Zhu et al.
BioTechniques, 68(3), 122-129 (2019-12-21)
Despite the controversy regarding the existence and physiological relevance of class A G protein-coupled receptor dimerization, there is substantial evidence for functional interactions between the dopamine D2 receptor (D2R) and the adenosine A2A receptor (A2AR). A2AR-D2R complexes have been detected in
Andry Andrianarivelo et al.
Science advances, 7(43), eabg5970-eabg5970 (2021-10-21)
[Figure: see text].
Ying Zhu et al.
Current protocols in neuroscience, 91(1), e86-e86 (2020-01-17)
Combining immunological and molecular biological methods, the antibody-based proximity ligation assay (PLA) has been used for more than a decade to detect and quantify protein-protein interactions, protein modification, and protein expression in situ, including in brain tissue. However, the transfer
Benjamin Lassus et al.
Scientific reports, 8(1), 17461-17461 (2018-12-01)
Although the prefrontal cortex and basal ganglia are functionally interconnected by parallel loops, cellular substrates underlying their interaction remain poorly understood. One novel approach for addressing this issue is microfluidics, a methodology which recapitulates several intrinsic and synaptic properties of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service