Skip to Content
Merck
All Photos(1)

Key Documents

8.43705

Sigma-Aldrich

Sodium triacetoxyborohydride

for synthesis

Synonym(s):

Sodium triacetoxyborohydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10BNaO6
CAS Number:
Molecular Weight:
211.94
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.22

Quality Level

form

powder

mp

110-114 °C

storage temp.

2-30°C

SMILES string

[Na+].[B-H](OC(=O)C)(OC(=O)C)OC(=O)C

InChI

1S/C6H10BO6.Na/c1-4(8)11-7(12-5(2)9)13-6(3)10;/h7H,1-3H3;/q-1;+1

InChI key

HHYFEYBWNZJVFQ-UHFFFAOYSA-N

Application

  • Chemical Synthesis: Used in the synthesis of (Piperazine-yl) Methanone derivatives with potential anticancer and antituberculosis activities, highlighting its role in facilitating N-alkylation reactions (Mallikarjuna et al., 2014).
  • Green Chemistry: Featured in the development of efficient N-methylation of secondary amines under solvent-free conditions, demonstrating its utility in sustainable chemical processes (Walter et al., 2024).
  • Organic Chemistry: Utilized in N-alkylation using carboxylic acids as alkyl sources, providing a versatile method for producing complex organic molecules (Tamura et al., 2018).

Analysis Note

Assay (by gasvolumetry): ≥ 96.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1 - Repr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken-Ichi Nakashima et al.
Journal of natural products, 82(12), 3347-3356 (2019-12-10)
Paraconiothins A-J (1-10), 10 new sesquiterpenoids, as well as five known sesquiterpenoids, were isolated from the cultures of the endophytic fungus Paraconiothyrium brasiliense ECN-258. The structures of the sesquiterpenoids were elucidated by extensive spectroscopic analysis. Furthermore, the absolute structures of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service