Skip to Content
Merck
All Photos(1)

Key Documents

8.20044

Sigma-Aldrich

2-Ethyltoluene

for synthesis

Synonym(s):

2-Ethyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5)C6H4CH3
CAS Number:
Molecular Weight:
120.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
210-255-1
NACRES:
NA.22

vapor pressure

2.8 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

440 °C

potency

5000 mg/kg LD50, oral (Rat)

bp

164-165 °C/1013 hPa

mp

-17 °C

transition temp

flash point 53 °C

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(C)c1c(cccc1)C

InChI

1S/C9H12/c1-3-9-7-5-4-6-8(9)2/h4-7H,3H2,1-2H3

InChI key

HYFLWBNQFMXCPA-UHFFFAOYSA-N

General description

2-Ethyltoluene also known as 1-Ethyl-2-methylbenzene, is an aromatic hydrocarbon, belongs to the group of alkyl-substituted benzene compounds. It is a non-polar solvent, making it suitable for dissolving non-polar and hydrophobic compounds. It is used as a building block in the synthesis of various organic compounds.

Specifications

Saturation Concentration in Air: 14 g/m3 (20 °C)

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.880 - 0.881
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service