Skip to Content
Merck
All Photos(1)

Key Documents

558255

Millipore

Saponin

A glycoside used to permeabilize cellular membranes. Exhibits hemolytic activity.

Synonym(s):

Saponin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 8365

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤10% ash

color

tan

solubility

water: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@H]2[C@@H](O[C@@H]([C@H]([C@@H]2O)O[C@@H]8O[C@@H]([C@H]([C@@H]([C@H]8O)O)O)CO)C(=O)O)O[C@@H]3[C@@](C4[C@@](C5[C@]([C@@]6(C[C@H]([C@]7([C@H]([C@@H](C(CC7C6=CC5)(C)C)OC(=O)\C(=C/C)\C)OC(=O)C)CO)O)C)(CC4)C)(CC3)C

InChI

1S/C55H86O24/c1-10-23(2)46(71)79-43-44(72-24(3)60)55(22-59)26(17-50(43,4)5)25-11-12-30-51(6)15-14-32(52(7,21-58)29(51)13-16-53(30,8)54(25,9)18-31(55)61)75-49-41(77-48-38(67)36(65)34(63)28(20-57)74-48)39(68)40(42(78-49)45(69)70)76-47-37(66)35(64)33(62)27(1

InChI key

AXNVHPCVMSNXNP-ZELRDNAQSA-N

General description

A glycoside used to permeabilize cellular membranes. Exhibits hemolytic activity.

Application


  • Complete biosynthesis of QS-21 in engineered yeast.: This article reports the successful biosynthesis of QS-21, a potent saponin adjuvant, in engineered yeast. The research highlights the potential for scalable and sustainable production of QS-21, which is crucial for vaccine development and immunotherapy. (Liu et al., 2024).


Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hi All, please, is this product suitable to detect bone marrow cell cycle in mice? Regards, Ahmad

    1 answer
    1. This product has not been tested for suitability to detect bone marrow cell cycle in mice. This is an application that will need to be determined by the end-user.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service