Skip to Content
Merck
All Photos(1)

Key Documents

234192-M

Millipore

Anti-Collagen Type VII Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, marmoset

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... COL7A1(1294)

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes type VII collagen.
Recognizes type VII collagen. Does not cross-react with fibronectin or type I, type II, type III, type IV, or type VI collagen.
This Anti-Collagen Type VII Rabbit pAb is validated for use in Frozen Sections, Immunoblotting, Immunofluorescence, Immunoprecipitation for the detection of Collagen Type VII.

Immunogen

Human
purified, human placenta collagen type VII

Application

Frozen Sections (1:20-1:60, see comments)

Immunoblotting (1:500-1:2000)

Immunofluorescence (1:200)

Immunoprecipitation (1:40-1:100)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Chen, M., et al. 1997. Invest. Dermatol. 108, 68.
Hashimoto, T., et al. 1996. Br. J. Dermatol. 134, 336.
Yasaka, N., et al. 1994. J. Dermatol. Sci. 7, 136.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service