Skip to Content
Merck
All Photos(2)

Key Documents

16-225

Sigma-Aldrich

Anti-Myc Tag Antibody, clone 4A6, Alexa Fluor 555 conjugate

clone 4A6, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
conjugate:
ALEXA FLUOR 555
application:
ICC
IF
WB
clone:
4A6, monoclonal
species reactivity:
human
citations:
3
technique(s):
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody product type

primary antibodies

clone

4A6, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

Specificity

Myc epitope tag (EQKLISEEDL)

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc. Clone 4A6

Application

Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
This Anti-Myc Tag Antibody, clone 4A6, Alexa Fluor 555 conjugate is validated for use in IF, WB, IC for the detection of Myc Tag.

Quality

routinely evaluated by immunoblot on RIPA lysates from transfected HeLa cells.

Target description

varies

Physical form

Protein G Purified

Storage and Stability

stable 1 year at 4°C from date of shipment

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rémi-Xavier Coux et al.
Development (Cambridge, England), 145(7) (2018-03-08)
Maintenance of cellular identity is essential for tissue development and homeostasis. At the molecular level, cell identity is determined by the coordinated activation and repression of defined sets of genes. The tumor suppressor L(3)mbt has been shown to secure cellular
Michelle G LeBlanc et al.
The Journal of cell biology, 216(9), 2945-2958 (2017-07-09)
The migration of primordial germ cells (PGCs) from their place of origin to the embryonic gonad is an essential reproductive feature in many animal species. In Drosophila melanogaster, a single G protein-coupled receptor, Trapped in endoderm 1 (Tre1), mediates germ
Overview of tag protein fusions: from molecular and biochemical fundamentals to commercial systems
Terpe, K
Applied Microbiology and Biotechnology, 60, 523-533 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service