Skip to Content
Merck
All Photos(1)

Documents

1.51487

Supelco

Pre-Column holder for HPLC Columns

for direct coupling of LiChroCART® 4-4 pre-Columns with Hibar® RT Columns

Synonym(s):

pre-column holder for TM=["LICHROCART"] guard cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

Pre-column holder for direct coupling of LiChroCART® 4-4 pre-columns with Hibar® RT columns,

parameter

250 bar pressure

technique(s)

HPLC: suitable

separation technique

supercritical fluid (SFC)

storage temp.

no temp limit

General description

This precolumn holder is suitable for direct connection to Hibar® HPLC columns.

Analysis Note

Extent of testing: passes test

Legal Information

Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service