Skip to Content
Merck
All Photos(1)

Key Documents

1.14549

Supelco

Iron Test, photometric

photometric, 0.05-4.00 mg/L (Fe), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Iron Cell Test, photometric, 0.05-4.00 mg/L (Fe), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

iron

measuring range

0.05-4.00 mg/L (Fe)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Triazine)

storage temp.

15-25°C

General description

Method: photometric 0.05 - 4.00 mg/l Fe Spectroquant®
This Spectroquant® Iron test allows the accurate quantification of iron ions in various waters. All iron ions are reduced to iron(II) ions by ascorbic acid.
In a thioglycolate-buffered medium these react with a triazine derivative to form a red-violet complex that is determined photometrically. MQuant® test strips can be used for pre-testing a sample in an economic and quick way to estimate the iron concentration and a possible dilution factor prior to using a quantitative method such as the Spectroquant® test.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination subsequent to acid mineralisation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service