1.12321
Samarium(III) oxide
99+
Synonym(s):
Samarium(III) oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
O3Sm2
CAS Number:
Molecular Weight:
348.72
MDL number:
UNSPSC Code:
12352303
EC Index Number:
235-043-6
NACRES:
NA.22
Recommended Products
Quality Level
form
solid
potency
>5000 mg/kg LD50, oral (Rat)
pH
8.0 (25 °C, 10 g/L in H2O, slurry)
mp
2325 °C
density
8.347 g/cm3
bulk density
850 kg/m3
storage temp.
no temp limit
SMILES string
[Sm+3].[Sm+3].[O-2].[O-2].[O-2]
InChI
1S/3O.2Sm/q3*-2;2*+3
InChI key
FKTOIHSPIPYAPE-UHFFFAOYSA-N
Analysis Note
Assay (complexometric, calc. on the ignited basis): ≥ 99.0 %
Loss on ignition: ≤ 2.0 %
Identity (ICP-OES): passes test
Loss on ignition: ≤ 2.0 %
Identity (ICP-OES): passes test
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service