Skip to Content
Merck
All Photos(1)

Key Documents

1.11142

Supelco

Residual Hardness Test Kit, colorimetric

water hardness 0.04-0.15 °d, water hardness 0.05-0.19 °e, 0.7-2.7 mg/L (CaCO₃), MQuant®

Synonym(s):

Residual Hardness Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Residual Hardness Test, colorimetric, water hardness 0.04-0.15 °d, water hardness 0.05-0.19 °e, 0.7-2.7 mg/L (CaCO3), for use with MQUANT®

specific analyte(s)

calcium, magnesium

Quality Level

measuring range

0.7-2.7 mg/L (CaCO3)

water hardness measuring range

0.04-0.15 °d
0.05-0.19 °e

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card 0.04 - 0.08 - 0.15 °d 0.05 - 0.1 - 0.19°e 0.7 - 1.4 - 2.7 mg/l CaCO₃ MQuant®
This MQuant® Residual Hardness test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card. Calcium and magnesium ions react with an indicator to form a red complex compound. The residual hardness is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card.
The test can be used in:
  • Boiler water and boiler feed water
  • Check of ion-exchanger plants for the production of deionized water

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service