1.07098
8-Hydroxyquinoline
GR for analysis Reag. Ph Eur
Synonym(s):
8-Hydroxyquinoline, Oxine, 8-Quinolinol
About This Item
Recommended Products
Agency
reag. Ph. Eur.
Quality Level
form
solid
potency
1200 mg/kg LD50, oral (Rat)
bp
267 °C/1013 hPa
mp
72.5-74.0 °C
density
1.034 g/cm3 at 20 °C
storage temp.
15-25°C
SMILES string
n1c2c(ccc1)cccc2O
InChI
1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H
InChI key
MCJGNVYPOGVAJF-UHFFFAOYSA-N
Application
- Pseudo-water-soluble Fe(2)O(3) as Nanozyme catalyzed chemiluminescent reaction for detection of brilliant blue in gelatin and beverages.: This innovative study utilizes 8-Hydroxyquinoline for enhancing chemiluminescent reactions in the detection of food colorants, presenting a new avenue in food safety and quality assurance techniques (Jabbar HS, 2024).
- Enhanced selectivity towards melanoma cells with zinc(II)-Schiff bases containing imidazole derivatives.: Incorporating 8-Hydroxyquinoline in the development of zinc(II)-Schiff base complexes, this research demonstrates potential therapeutic applications in targeting melanoma cells, highlighting its role in medicinal chemistry (Côrte-Real et al., 2024).
- Synthesis, crystal structure and anti-cancer activity of the complex chlorido-(η(2)-ethyl-ene)(quinolin-8-olato-κ(2)N,O)platinum(II) by experimental and theoretical methods.: This paper details the synthesis and anti-cancer properties of a novel platinum(II) complex using 8-Hydroxyquinoline, offering insights into its structural characteristics and therapeutic potential (Chi NTT et al., 2024).
- Anticancer activity of 8-hydroxyquinoline-triphenylphosphine rhodium(III) complexes targeting mitophagy pathways.: Exploring the use of 8-Hydroxyquinoline in rhodium(III) complexes, this study provides a new perspective on targeting cancer cell metabolism, specifically through mitophagy pathways (Huang XQ et al., 2024).
- Mitochondria-localizing triphenylphosphine-8-hydroxyquinoline Ru complexes induce ferroptosis and their antitumor evaluation.: This research examines the effectiveness of 8-Hydroxyquinoline complexes in inducing ferroptosis in cancer cells, a novel approach in cancer therapy focusing on oxidative cell death mechanisms (Meng T et al., 2024).
Analysis Note
Identity (IR-spectrum): passes test
Matter insoluble in ethanol: ≤ 0.05 %
Melting point: 73 - 75 °C
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.01 %
Sulfated ash: ≤ 0.05 %
Suitability as reagent (for Mg-test): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service