Skip to Content
Merck
All Photos(1)

Key Documents

1.06450

Supelco

Sodium fluoride

99.99 Suprapur®

Synonym(s):

Sodium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FNa
CAS Number:
Molecular Weight:
41.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-667-8
NACRES:
NB.21
Assay:
99.99%
grade:
for inorganic trace analysis
form:
crystals

grade

for inorganic trace analysis

Quality Level

vapor pressure

1 hPa ( 1077 °C)

Assay

99.99%

form

crystals

potency

148.5 mg/kg LD50, oral (Rat)

pH

10.2 (20 °C, 40 g/L in H2O)

mp

996 °C

density

2.8 g/cm3 at 20 °C

cation traces

Ba: ≤5 ppm
Ca: ≤5 ppm
Co: ≤0.05 ppm
Cu: ≤0.05 ppm
Fe: ≤0.5 ppm
K: <2 ppm
Li: ≤0.5 ppm
Mg: ≤1 ppm
Mn: ≤0.05 ppm
Ni: ≤0.05 ppm
Pb: ≤0.1 ppm
Sr: ≤1 ppm
Zn: ≤0.2 ppm

storage temp.

2-30°C

SMILES string

[Na+].[F-]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Application


  • Investigations of the reaction mechanism of sodium with hydrogen fluoride to form sodium fluoride and the adsorption of hydrogen fluoride on sodium fluoride monomer and tetramer.: This paper delves into the reaction mechanisms between sodium and hydrogen fluoride, and the adsorption properties on sodium fluoride structures. These findings are essential for chemists working on chemical synthesis, reaction kinetics, and industrial applications of fluoride compounds (Yu et al., 2024).

Analysis Note

Purity (metallic): ≥ 99.99 %
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 5 ppm
Co (Cobalt): ≤ 0.05 ppm
Cu (Copper): ≤ 0.05 ppm
Fe (Iron): ≤ 0.5 ppm
K (Potassium): ≤ 2 ppm
Li (Lithium): ≤ 0.5 ppm
Mg (Magnesium): ≤ 1 ppm
Mn (Manganese): ≤ 0.05 ppm
Ni (Nickel): ≤ 0.05 ppm
Pb (Lead): ≤ 0.1 ppm
Sr (Strontium): ≤ 1 ppm
Zn (Zinc): ≤ 0.2 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service