Skip to Content
Merck

1.05683

SAFC

tri-Lithium citrate tetrahydrate

EMPROVE® ESSENTIAL Ph Eur,BP

Pharma Manufacturing

Synonym(s):

tri-Lithium citrate tetrahydrate, Trilithium citrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Li3O7 · 4H2O
CAS Number:
Molecular Weight:
281.98
MDL number:
UNSPSC Code:
12352107
EC Index Number:
213-045-8

Agency

BP
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

pH

8.5 (25 °C, 50 g/L in H2O)

mp

105 °C

bulk density

420 kg/m3

application(s)

pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[Li+].[Li+].[Li+].[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-].O.O.O.O

InChI

1S/C6H8O7.3Li.4H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;4*1H2/q;3*+1;;;;/p-3

InChI key

HXGWMCJZLNWEBC-UHFFFAOYSA-K

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The substance 1.05683 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service