Skip to Content
Merck
All Photos(1)

Key Documents

1.01846

Supelco

Manganese Test, photometric

photometric, 0.005-2.00 mg/L (Mn), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Manganese Test, photometric, 0.005-2.00 mg/L (Mn), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 250 tests

specific analyte(s)

manganese

measuring range

0.005-2.00 mg/L (Mn)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (PAN)

storage temp.

15-25°C

General description

Method: photometric 0.005 - 2.00 mg/l Mn Spectroquant®
This Spectroquant® Manganese Reagent Test allows the accurate quantification of the concentration of all manganese ion species in aqueous samples.
Method applied: All manganese ions are reduced to manganese(II) ions using ascorbic acid. In weakly alkaline solution these to manganese(II) ions react with 1-(2-pyridylazo)-2-naphthol (PAN) to form a red complex which is determined photometrically.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service