Skip to Content
Merck
All Photos(1)

Key Documents

1.00229

Supelco

Chromium(VI) oxide

for analysis EMSURE®

Synonym(s):

Chromium(VI) oxide, Chromic anhydride, Chromium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-607-8
NACRES:
NA.21
form:
solid
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥99.0% (iodometric)

form

solid

impurities

≤0.02% Substances precipitated by ammonia

loss

≤ 1.0% loss on drying, 105°C

pH

<1 (20 °C, 100 g/L in H2O)

mp

197 °C

density

2.7 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.01%
K: ≤0.005%
N: ≤0.005%
Na: ≤0.2%

storage temp.

2-30°C

SMILES string

[Cr](=O)(=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (iodometric): ≥ 99.0 %
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.02 %
Total nitrogen (N): ≤ 0.005 %
Fe (Iron): ≤ 0.01 %
K (Potassium): ≤ 0.005 %
Na (Sodium): ≤ 0.2 %
Substances precipitated by ammonia: ≤ 0.02 %
Loss on Drying (105 °C): ≤ 1.0 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service