N-920
Norbuprenorphine-D3 solution
100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
Norbuprenorphine-d3
About This Item
grade
certified reference material
Quality Level
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
Narcotic Licence Schedule A (Switzerland)
concentration
100 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
[2H]C([2H])([2H])O[C@]12CC[C@@]3(C[C@@H]1[C@](C)(O)C(C)(C)C)[C@H]4Cc5ccc(O)c6O[C@@H]2[C@]3(CCN4)c56
InChI
1S/C25H35NO4/c1-21(2,3)22(4,28)16-13-23-8-9-25(16,29-5)20-24(23)10-11-26-17(23)12-14-6-7-15(27)19(30-20)18(14)24/h6-7,16-17,20,26-28H,8-13H2,1-5H3/t16-,17-,20-,22+,23-,24+,25-/m1/s1/i5D3
InChI key
YOYLLRBMGQRFTN-QLWKBOQPSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Improving GC-MS Analysis of Norbuprenorphine: A study by Gervais JR and Hobbs GA discusses the use of an acetyl derivative to enhance the GC-MS analysis of Norbuprenorphine in urine, particularly in the presence of high concentrations of Buprenorphine, highlighting its significance as an analytical standard in forensic and clinical toxicology (Gervais and Hobbs, 2016).
- Quantification of Acetaminophen Protein Adducts: Cook SF et al. developed a method for the quantification of a biomarker for acetaminophen protein adducts in human serum using high-performance liquid chromatography-electrospray ionization-tandem mass spectrometry. This study demonstrates the application of advanced analytical techniques in toxicology and pharmacokinetics, where Norbuprenorphine-D3 serves as a reliable internal standard (Cook et al., 2015).
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service