H-176
5-Hydroxyindole-3-acetic acid (5-HIAA) solution
1 mg/mL ( Degassed Methanol w/ 0.1% BHT), certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
5-HIAA, 5-Hydroxyindoleacetic acid
About This Item
Recommended Products
grade
certified reference material
Quality Level
description
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1 mg/mL ( Degassed Methanol w/ 0.1% BHT)
format
single component solution
storage temp.
-65 to -96°C
SMILES string
[nH]1c2c(c(c1)CC(=O)O)cc(cc2)O
InChI key
DUUGKQCEGZLZNO-UHFFFAOYSA-N
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F
Flash Point(C)
9.7 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service