Skip to Content
Merck
All Photos(1)

Documents

D-046

Supelco

Dehydronorketamine hydrochloride solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12ClNO · HCl
Molecular Weight:
258.14
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C1C(C2=C(Cl)C=CC=C2)(N)CCC=C1.[H]Cl

InChI

1S/C12H12ClNO.ClH/c13-10-6-2-1-5-9(10)12(14)8-4-3-7-11(12)15;/h1-3,5-7H,4,8,14H2;1H

InChI key

WQWIJCRGQUZVJW-UHFFFAOYSA-N

General description

Dehydronorketamine, also known as DHNK, is a secondary metabolite of the anesthetic drug ketamine. Ketamine is widely used in human and veterinary medicine for the induction and maintenance of general anesthesia and at parties or raves as a noted dissociative club drug. This certified Snap-N-Spike® solution is suitable for numerous GC/MS or LC/MS testing methods in forensic analysis, clinical toxicology, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service