Skip to Content
Merck
All Photos(1)

Key Documents

860531P

Avanti

C12 Ceramide-1-Phosphate (d18:1/12:0)

Avanti Research - A Croda Brand 860531P, powder

Synonym(s):

N-lauroyl-ceramide-1-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H63N2O6P
CAS Number:
Molecular Weight:
578.80
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860531P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860531P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCC)=O)COP([O-])(O)=O.[NH4+]

InChI

1S/C30H60NO6P.H3N/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-29(32)28(27-37-38(34,35)36)31-30(33)26-24-22-20-17-12-10-8-6-4-2;/h23,25,28-29,32H,3-22,24,26-27H2,1-2H3,(H,31,33)(H2,34,35,36);1H3/b25-23+;/t28-,29+;/m0./s1

InChI key

BZXZJNUBNUWPEQ-GMVBIGKRSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (860531P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Gómez-Muñoz et al.
Journal of lipid research, 45(1), 99-105 (2003-10-03)
It was reported previously that ceramide-1-phosphate (Cer-1-P) is mitogenic for fibroblasts (Gómez-Muñoz, A., P. A. Duffy, A. Martin, L. O'Brien, H-S. Byun, R. Bittman, and D. N. Brindley. 1995. Mol. Pharmacol. 47: 883-889; Gómez-Muñoz, A., L. M. Frago, L. Alvarez
A Gomez-Muñoz et al.
Molecular pharmacology, 47(5), 833-839 (1995-05-01)
Ceramide and ceramide-1-phosphate are sphingolipid analogues of diacylglycerol and phosphatidate, respectively, and they are putative second messengers of agonist-stimulated sphingomyelin metabolism. The interactions of exogenous cell-permeable ceramides and ceramide-1-phosphates in modifying DNA synthesis and signal transduction were investigated in Rat-1
V T Hinkovska-Galcheva et al.
The Journal of biological chemistry, 273(50), 33203-33209 (1998-12-05)
Ceramide, a product of agonist-stimulated sphingomyelinase activation, is known to be generated during the phagocytosis of antibody-coated erythrocytes by polymorphonuclear leukocytes. Agonist-stimulated formation of ceramide-1-phosphate is now shown to occur in 32PO4-labeled neutrophils. Ceramide-1-phosphate is formed by a calcium-dependent ceramide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service