Skip to Content
Merck
All Photos(2)

Key Documents

860203O

Avanti

18:1 Phosphatidylbutanol

1,2-dioleoyl-sn-glycero-3-phosphobutanol (sodium salt), neat oil

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphobutanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H80O8PNa
CAS Number:
Molecular Weight:
779.05
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 25 mg (860203O-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860203O

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C43H81O8P.Na/c1-4-7-10-12-14-16-18-20-22-24-26-28-30-32-34-36-42(44)48-39-41(40-50-52(46,47)49-38-9-6-3)51-43(45)37-35-33-31-29-27-25-23-21-19-17-15-13-11-8-5-2;/h20-23,41H,4-19,24-40H2,1-3H3,(H,46,47);/q;+1/p-1/b22-20-,23-21-;/t41-;/m1./s1

InChI key

BXQFBLMPDPPGFW-RLWYHRQISA-M

Application

18:1 Phosphatidylbutanol or 1,2-dioleoyl-sn-glycero-3-phosphobutanol (sodium salt) may be used:
  • to monitor the changes in phosphatidic acid (PA) dynamics in pollen tube after pharmacological treatments
  • as an internal standard for extraction of phospholipids from whole blood
  • as a standard to measure phospholipase D (PLD) activity in rat 2 fibroblasts by thin layer chromatograpghy (TLC)

Packaging

5 mL Clear Glass Sealed Ampule (860203O-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Nalesso et al.
Electrophoresis, 31(7), 1227-1233 (2010-03-03)
Phosphatidylethanol (Peth), a group of aberrant phospholipids formed in cell membranes in the presence of ethanol, has been recently proposed as biomarker of chronic alcohol abuse. The aim of this study was to develop a new analytical method, based on
Martin Potocký et al.
The New phytologist, 203(2), 483-494 (2014-04-23)
Although phosphatidic acid (PA) is structurally the simplest membrane phospholipid, it has been implicated in the regulation of many cellular events, including cytoskeletal dynamics, membrane trafficking and stress responses. Plant PA shows rapid turnover but the information about its spatio-temporal
Jeremiah N Winter et al.
American journal of physiology. Cell physiology, 299(2), C335-C344 (2010-04-30)
The mammalian target of rapamycin (mTOR) assembles into two distinct multiprotein complexes known as mTORC1 and mTORC2. Of the two complexes, mTORC1 acts to integrate a variety of positive and negative signals to downstream targets that regulate cell growth. The
Maria A Frias et al.
The Journal of biological chemistry, 295(1), 263-274 (2019-11-27)
Mammalian target of rapamycin complex 1 (mTORC1) promotes cell growth and proliferation in response to nutrients and growth factors. Amino acids induce lysosomal translocation of mTORC1 via the Rag GTPases. Growth factors activate Ras homolog enriched in brain (Rheb), which
Y Liu et al.
American journal of physiology. Lung cellular and molecular physiology, 295(3), L471-L478 (2008-07-16)
We have previously reported the participation of mitogen-activated protein, Rho, and phosphoinositide-3 (PI3) kinases in separate pathways in serotonin (5-HT)-induced proliferation of pulmonary artery smooth muscle cells (SMCs). In this study, we investigated the possible participation of phospholipase D (PLD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service