Skip to Content
Merck
All Photos(2)

Key Documents

857137P

Avanti

18:1 BDP (S,S)

sn-[2,3-dioleoyl]-glycerol-1-phospho-sn-1′-[2′,3′-dioleoyl]-glycerol (ammonium salt), powder

Synonym(s):

110858

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H146NO12P
CAS Number:
Molecular Weight:
1320.97
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (857137P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857137P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](COP(OC[C@@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)([O-])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C78H143O12P.H3N/c1-5-9-13-17-21-25-29-33-37-41-45-49-53-57-61-65-75(79)85-69-73(89-77(81)67-63-59-55-51-47-43-39-35-31-27-23-19-15-11-7-3)71-87-91(83,84)88-72-74(90-78(82)68-64-60-56-52-48-44-40-36-32-28-24-20-16-12-8-4)70-86-76(80)66-62-58-54-50-46-42

InChI key

JKVJJSANRWKSFX-GBKYCRPNSA-N

Packaging

5 mL Clear Glass Sealed Ampule (857137P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service