Skip to Content
Merck
All Photos(2)

Key Documents

840485C

Avanti

18:2 PG

1,2-dilinoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1,2-di-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(18:2(9Z,12Z)/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H74O10PNa
CAS Number:
Molecular Weight:
792.99
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840485C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840485C

concentration

10 mg/mL (840485C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCC\C=C/C\C=C/CCCCC)OCC(O)CO

InChI

1S/C42H75O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h11-14,17-20,39-40,43-44H,3-10,15-16,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b13-11-,14-12-,19-17-,20-18-;/t39?,40-;/m1./s1

InChI key

ZIOOVFPFWVCCEB-WDFQPZSRSA-M

General description

18:2 PG or 1,2-dilinoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) is a sodium salt of glycerophospholipid, with substitution of two chains of linolenic acid at the sn-1 and sn-2 positions and phospho-(1′-rac-glycerol) at the sn-3 position of the glycerol backbone. It is a poly-unsaturated lipid molecule.

Application

18:2 PG or 1,2-dilinoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt) might be used in saposin-derived lipid nanoparticles (SapNPs) formation to study the lipid specificity of saposins.

Packaging

5 mL Clear Glass Sealed Ampule (840485C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Flayhan et al.
Structure (London, England : 1993), 26(2), 345-355 (2018-02-08)
Saposin-derived lipid nanoparticles (SapNPs) are a new alternative tool for membrane protein reconstitution. Here we demonstrate the potential and advantages of SapNPs. We show that SapA has the lowest lipid specificity for SapNP formation. These nanoparticles are modular and offer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service