Skip to Content
Merck
All Photos(2)

Key Documents

810143P

Avanti

14:0 NBD PE

Avanti Research - A Croda Brand 810143P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H70N5O11P
CAS Number:
Molecular Weight:
815.97
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810143P-1mg)
pkg of 5 × 1 mg (810143P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810143P

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is a fluorescently labeled lipid localized at the membrane interface. Phosphoethanolamine is the second most abundant, metabolically related aminophospholipid. It is mostly enriched in brain.

Application

14:0 NBD PE may be used in the lipid mixing assay for the preparation of phospholipid vesicles and to study its non-equilibrium and equilibrium variations in thermodynamic state of the lipid-water-interface

Biochem/physiol Actions

Phosphoethanolamine of cardiac myocytes is known to prevent cellular damage after ischemia. N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) contributes to the Red edge excitation shift (REES) effect monitoring because of its unique motional and dielectric properties.

Packaging

5 mL Amber Glass Screw Cap Vial (810143P-1mg)
5 mL Amber Glass Screw Cap Vial (810143P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shamit Shrivastava et al.
PloS one, 8(7), e67524-e67524 (2013-07-19)
Fluorescent dyes are vital for studying static and dynamic patterns and pattern formation in cell biology. Emission properties of the dyes incorporated in a biological interface are known to be sensitive to their local environment. We report that the fluorescence
In vitro Membrane Interaction and Liposome Fusion Assays Using Recombinant Hepatitis C Virus Envelope Protein E2
Shrivastava S and Schneider MF
Bio-protocol, 8(23), 543-554 (2018)
Application of NBD-labeled lipids in membrane and cell biology
Fluorescent methods to study biological membranes, 37-50 (2012)
Jean E Vance et al.
Biochimica et biophysica acta, 1831(3), 543-554 (2012-09-11)
Phosphatidylserine (PS) and phosphatidylethanolamine (PE) are metabolically related membrane aminophospholipids. In mammalian cells, PS is required for targeting and function of several intracellular signaling proteins. Moreover, PS is asymmetrically distributed in the plasma membrane. Although PS is highly enriched in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service