Skip to Content
Merck
All Photos(3)

Key Documents

Z744083

Sigma-Aldrich

H-Genie® Lite

Synonym(s):

H-Genie® Lite High-Pressure H2 Generator, H-Genie® Lite Smart Hydrogen Generator, H-Genie® Lite Hydrogen Generator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111604
NACRES:
NA.22

AC/DC input

100 - 240 V AC, 50 - 60 Hz (Plug types: Type B, Type C, Type J, Type G)

parameter

(Hydrogen Production Rate: 1 NL/min)
(Output Pressure Range: 1-50 bar)
(Recommended environment: Ventilated laboratory fume hood)
(Water consumption rate: 200 mL/hr)
(Water requirements: Deionized water (recommended conductivity < 1µS/cm))
(Water reservoir capacity: 3L)

unit H × W × D

385 mm × 365 mm × 476 mm

weight

33.7 kg (unit)

fitting

1/8 in. Swagelok outlet tube (stainless steel)

General description

It is a benchtop hydrogen generator for use in batch processes. Producing gaseous hydrogen from water up to 50 bar (725 psi) on-demand, it is an easy and safe solution to replace hydrogen cylinders and minimize H2 gas hazards on site. This compact laboratory hydrogen generator produces 3.0 purity hydrogen from water at pressures from 1-50 bar at a flow rate of 1 L/min.

Ideal for seamless hydrogen-based experiments with multiple batch systems, the set-up of this high-pressure smart hydrogen generator is as simple as click and go.

H-Genie® Lite uses H-Genie® Lite water purifier cartridges (Z744084) for hydrogen generation.

Application

H2 generator designed for use with balloons, Parr shakers, and pressure reactors.

Features and Benefits

  • Hydrogen gas safety: Equipped with safety features, such as built-in internal hydrogen leak detectors.
  • Ease of use: Capable of generating hydrogen from deionized water, with a simple click-and-go set-up. With a touchscreen display, all you need to do is set the required pressure or time and press start. Simplify your hydrogenation process, as it is compatible with any reactors and balloons; in addition, you can run multiple reactors with one H-Genie® Lite.
  • Compact hydrogen generator: Acts as a space-saving alternative. No longer worry about hydrogen cylinders in the lab as this high-purity hydrogen gas generator can easily be moved between fume hoods.

Legal Information

H-Genie is a registered trademark of ThalesNano Energy

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service