Z741012
Aldrich® Essentials round media storage bottle
wide mouth, capacity 250 mL, GL 45 polypropylene cap with pour ring
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
borosilicate glass bottle
polypropylene cap
round bottle
Joint
Joints threaded neck (GL45)
sterility
non-sterile
feature
graduations
with cap
packaging
pkg of 10 ea
manufacturer/tradename
Sigma-Aldrich
capacity
250 mL
Related Categories
General description
Borosilicate glass, wide mouth, media/reagent bottles with polypropylene GL 45 thread screw cap with pour ring. Large marking spot and reference graduations.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)