Key Documents
Z284270
Spinbar® Pyrex® magnetic stir bars
size 1 1/2 in. × 3/8 in.
Synonym(s):
Stirbar, Stirring Bar, glass Stir Bar
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Pricing and availability is not currently available.
material
cylindrical
round glass coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37101-0112
L
38 mm
diam.
9.5 mm
size
1 1/2 in. × 3/8 in.
General description
  • Spinbar® Pyrex® magnetic stir bars are manufactured from Type I borosilicate glass.
  • Ideal for use in applications requiring high temperature up to 274°C (525°F).
  • The smooth Pyrex®glass coating ensures easy cleaning and offers “zero absorption” thus preventing cross-contamination of samples.
  • Autoclavable.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)