Z114065
Wheaton glass serum bottle
volume 100 mL, amber, O.D. 20 mm, diam. × H 52 mm × 95 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
borosilicate glass bottle
description
USP Type I
feature
autoclavable
closure type crimp top vial
O.D.
20 mm
capacity
100 mL
diam. × H
52 mm × 95 mm
volume
100 mL
color
amber
fitting
opening for 20 mm
Looking for similar products? Visit Product Comparison Guide
General description
It is autoclavable, easy sterilization, is used to contain substances to prevent contamination.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service