Key Documents
Z102431
Double-ended needle
gauge 16, L 24 in.
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science
L
24 in.
gauge
16
Related Categories
General description
304 SS. One noncoring tip, one flat-cut end.
Choose from one of the most recent versions:
Certificates of Analysis (COA)