Skip to Content
Merck
All Photos(1)

Key Documents

W363499

Sigma-Aldrich

4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one

natural, ≥97%, FG

Synonym(s):

Caramel furanone, Sotolon, Sugar lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
FEMA Number:
3634
MDL number:
UNSPSC Code:
12164502
Flavis number:
10.030
NACRES:
NA.21
Organoleptic:
caramel; maple; sweet
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
sugar cane
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

sugar cane

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.491 (lit.)

bp

184 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; maple; sweet

SMILES string

CC1OC(=O)C(O)=C1C

InChI

1S/C6H8O3/c1-3-4(2)9-6(8)5(3)7/h4,7H,1-2H3

InChI key

UNYNVICDCJHOPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service