THSD1143
ThalesNano MidiCart™ catalyst cartridge system
L × diam. 116 mm × 9.2 mm, 10% Pt/C
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pack of 1 ea
manufacturer/tradename
ThalesNano THS D1143
L × diam.
116 mm × 9.2 mm
Looking for similar products? Visit Product Comparison Guide
General description
MiDiCarts are suitable for use only with Thalesnano H-Cube MidiCart reactors.
Legal Information
MidiCart is a trademark of ThalesNano Nanotechnology Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service