SYNF31291L
Synthware™ recovery flask
1000 mL, joint: ST/NS 29/42
Synonym(s):
Synthware F31291L, oval shape flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F31291L
capacity
1,000 mL
1000 mL
1000 mL
Related Categories
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)