All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C144H285N2O59P
Molecular Weight:
3019.76
UNSPSC Code:
51171641
NACRES:
NA.22
Recommended Products
Assay
>90%
form
solid or viscous liquid
reaction suitability
reagent type: cross-linking reagent
reagent type: payload delivery reagent
functional group
phospholipid
polymer architecture
shape: linear
functionality: monofunctional
shipped in
ambient
storage temp.
−20°C
Related Categories
Features and Benefits
m-dPEG®25-amido-dPEG®25-DSPE is a micelle building block.
Legal Information
Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service