Skip to Content
Merck
All Photos(3)

Key Documents

P13002

Sigma-Aldrich

1,10-Phenanthroline monohydrochloride monohydrate

97%

Synonym(s):

1,10-Phenanthroline hydrochloride monohydrate, o-Phenanthroline hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2 · HCl · H2O
CAS Number:
Molecular Weight:
234.68
Beilstein:
4007967
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

224-225 °C (lit.)

SMILES string

O.Cl.c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2.ClH.H2O/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;/h1-8H;1H;1H2

InChI key

NDLHUHRGAIHALB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T M-H Nguyen et al.
International journal of pharmaceutics, 482(1-2), 21-26 (2014-12-03)
Hyperphosphatemia is one of the main risk factors contributing to morbidity and mortality in patients with end stage renal disease. The demand for a new phosphate binder is continuously increasing since the number of patients suffering under hyperphosphatemia is growing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service