MT51344760
Mettler Toledo Ionic Strength Adjuster
for Solid State Ion Selective Electrodes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115600
NACRES:
NB.79
Recommended Products
form
liquid
manufacturer/tradename
Mettler-Toledo 51344760
volume
475 mL
Looking for similar products? Visit Product Comparison Guide
General description
Ionic strength adjuster (ISA) provides a constant background ionic strength for samples and standards.
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service