KITALYSIS-24PD
KitAlysis™ High-Throughput 24 Pd Precatalyst Reaction Screening Kit-Pack of 2
Synonym(s):
Pd Precatalysts
About This Item
Recommended Products
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings
functional group
phosphine
1 of 4
This Item | KITALYSIS-PHO | KITALYSIS-BOR | 916455 |
---|---|---|---|
functional group phosphine | functional group - | functional group phosphine | functional group - |
Application
Step-by-Step Guide for KitAlysis High-Throughput Palladium Precatalyst Cross-Coupling Reaction Screening Kit
Downloadable experimental Excel Sheet
Each kit contains 2 customizable screening sets that include the following components:
- 24 pre-weighed catalysts in glass micro vials loaded with stir bars and topped with cap mat.
- 2 KitAlysis 24-Well reaction block replacement films.
- 4 empty substrate stock solution reaction vials with stir bars.
- 3 stir bars for stock solutions.
- 1 internal standard.
Monitor reaction progress via TLC-MS by following this protocol
Best when used with KitAlysis Benchtop Inertion Box and 24-well Reaction Block (sold separately).
Features and Benefits
- Designed and tested by synthetic chemists.
- Easily run 24-microscale reaction in parallel.
- Suggested conditions: 10 μmol reactions with 1 μmol catalyst (10 mol%) 0.1 M concentration, and 3 equiv. base.
- Customizable and downloadable excel template for quick calculations and easy incorporation into electronic laboratory notebooks.
Legal Information
recommended
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Supplementary Hazards
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
KitAlysis High-Throughput Screening Kits provide solution to efficiently identify or optimize suitable catalytic reaction conditions. Chemist can rapidly run 24 unique micro scale reactions in parallel with tailored conditions.
Materials Included in your KITALYSIS-24PD-2PK High-Throughput Screening Kit
Protocols
Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.
Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.
Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.
Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.
Related Content
Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.
Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.
Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.
Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service