HAM59000-15
Hamilton® Microlab® 600 instrument syringe
model, 1705 BFP, volume 50 μL
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
material
Gastight Borosilicate glass/PTFE syringe
feature
model 1705 BFP
manufacturer/tradename
Hamilton 59000-15
parameter
10-115 °C temp. range (50-239 °F)
1000 psig pressure
I.D. × O.D.
1.03 mm × 7.75 mm
volume
50 μL
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)