GF71330767
Silver
foil, 25mm disks, thickness 0.007mm, 99.97%
Synonym(s):
Silver, AG000150
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
≥99.97%
form
foil
manufacturer/tradename
Goodfellow 713-307-67
resistivity
1.59 μΩ-cm, 20°C
diam. × thickness
25 mm × 0.007 mm
bp
2212 °C (lit.)
mp
960 °C (lit.)
density
10.49 g/cm3 (lit.)
SMILES string
[Ag]
InChI
1S/Ag
InChI key
BQCADISMDOOEFD-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Surface-enhanced Raman spectroscopy and density functional theory study on 4,4'-bipyridine molecule.
Zhiping Zhuang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(2), 509-516 (2006-09-22)
The molecular geometry and vibrational frequencies of 4,4'-bipyridine (BPE) in the ground state were calculated using density functional theory (DFT) methods (B3LYP) with 6-31++G(d,p) basis set. The optimized geometric bond lengths and bond angles are obtained by DFT employing the
H H Wiebe et al.
Plant physiology, 59(2), 256-258 (1977-02-01)
Water potential was monitored at nine locations along single maize (Zea mays L.) leaf blades with aluminum block in situ thermocouple hygrometers. Water potential showed a continuous decrease toward the tip, with a 2- to 4-bar difference between leaf base
Ken-ichiro Matsumoto et al.
Biological & pharmaceutical bulletin, 28(11), 2029-2034 (2005-11-08)
A novel screening experiment, to find radioactive probes for non-invasive measurements of physiological functions in experimental animals, was tested using the in vivo multitracer analysis technique. The details of the efficiency of the detector settings used in the in vivo
R D Stickrod et al.
Rapid communications in mass spectrometry : RCM, 14(14), 1266-1268 (2000-08-05)
Nitrate-delta(15)N from groundwater samples is determined on an inorganic nitrate derivative using automated, continuous-flow elemental analyzer/isotope ratio mass spectrometry (EA/IRMS). Nitrate is extracted and concentrated based on a recently published ion-exchange resin method. Freeze-dried AgNO(3) (0.5-1.5 mg) is packed in
M Dubiel et al.
Journal of synchrotron radiation, 8(Pt 2), 539-541 (2001-08-22)
The thermal expansion behaviour of silver fcc has been described by an anharmonic Einstein model using EXAFS data in the temperature range between 10 and 300 K. The linear expansion coefficient of a bulk silver foil agrees well with X-ray
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service