GF49763403
Havar® - high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn
foil, 150 x 150mm, 0.035mm thickness, as rolled, light tight (lt)
Synonym(s):
CO010260
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
light tight (lt)
form
foil (as rolled)
manufacturer/tradename
Goodfellow 497-634-03
size × thickness
150 mm × 150 mm × 0.035 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Havar is a registered trademark of Precision Metals, Inc. AMETEK, or its subsidiaries or affiliates.
Havar is a registered trademark of Precision Metals, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service