Skip to Content
Merck
All Photos(1)

Key Documents

F15323

Sigma-Aldrich

2-Fluorotoluene

≥99%

Synonym(s):

1-Fluoro-2-methylbenzene, o-Fluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4F
CAS Number:
Molecular Weight:
110.13
Beilstein:
1853362
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.8 (vs air)

Quality Level

vapor pressure

21 mmHg ( 20 °C)

Assay

≥99%

refractive index

n20/D 1.473 (lit.)

bp

113-114 °C (lit.)

mp

−62 °C (lit.)

density

1.001 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1F

InChI

1S/C7H7F/c1-6-4-2-3-5-7(6)8/h2-5H,1H3

InChI key

MMZYCBHLNZVROM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frans Everson et al.
International journal of environmental research and public health, 16(13) (2019-07-03)
Exposure to ambient NO2 and benzene, toluene ethyl-benzene and m+p- and o-xylenes (BTEX) is associated with adverse cardiovascular effects, but limited information is available on the effects of personal exposure to these compounds in South African populations. This 6-month follow-up
Frans Everson et al.
Environmental research, 182, 108993-108993 (2019-12-13)
Air pollution exposure is a major global health concern and has been associated with molecular aging. Unfortunately, the situation has not received much attention in the African region. The aim of this study was to investigate whether current personal ambient
Aurélien Rey et al.
Journal of chromatography. A, 1404, 95-103 (2015-06-09)
A new strategy was developed to elucidate and quantify 56 (69 analytes including isomers) suspected chemically defined fragrance allergens in perfumes that were recently targeted by the Scientific Committee on Consumer Safety (SCCS). Samples were analyzed with a two-dimensional gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service