Skip to Content
Merck
All Photos(1)

Documents

D42402

Sigma-Aldrich

1,9-Dibromononane

97%

Synonym(s):

Nonamethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)9Br
CAS Number:
Molecular Weight:
286.05
Beilstein:
1737598
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.496 (lit.)

bp

285-288 °C (lit.)

density

1.407 g/mL at 25 °C (lit.)

SMILES string

BrCCCCCCCCCBr

InChI

1S/C9H18Br2/c10-8-6-4-2-1-3-5-7-9-11/h1-9H2

InChI key

WGAXVZXBFBHLMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan-Ping Pang et al.
PloS one, 4(2), e4349-e4349 (2009-02-06)
Aphids, among the most destructive insects to world agriculture, are mainly controlled by organophosphate insecticides that disable the catalytic serine residue of acetylcholinesterase (AChE). Because these agents also affect vertebrate AChEs, they are toxic to non-target species including humans and
Rahul A Patil et al.
Analytical and bioanalytical chemistry, 410(19), 4645-4655 (2018-01-31)
The rapid expansion in the study and use of ionic liquids (ILs) is a result of their unique properties including negligible volatility, high thermal stability, and ability to dissolve disparate compounds. However, because ILs have infinitely variable structures (often referred
Mojca Seručnik et al.
The journal of physical chemistry. B, 122(21), 5381-5388 (2018-01-26)
Complexes of polycations and DNA, also known as polyplexes, have been extensively studied in the past decade, as potential gene delivery systems. Their stability depends strongly on the characteristics of the polycations, as well as the nature of the added

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service