C105104
2-Cyclohexylethanol
99%
Synonym(s):
(2-Hydroxyethyl)cyclohexane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H11CH2CH2OH
CAS Number:
Molecular Weight:
128.21
Beilstein:
1848152
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.465 (lit.)
bp
206-207 °C/745 mmHg (lit.)
density
0.919 g/mL at 25 °C (lit.)
SMILES string
OCCC1CCCCC1
InChI
1S/C8H16O/c9-7-6-8-4-2-1-3-5-8/h8-9H,1-7H2
InChI key
QJQZRLXDLORINA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
186.8 °F - closed cup
Flash Point(C)
86 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rabea Schlüter et al.
Applied microbiology and biotechnology, 103(10), 4137-4151 (2019-04-04)
The cycloalkanes, comprising up to 45% of the hydrocarbon fraction, occur in crude oil or refined oil products (e.g., gasoline) mainly as alkylated cyclohexane derivatives and have been increasingly found in environmental samples of soil and water. Furthermore, short-chain alkylated
Qinglei Meng et al.
Nature communications, 8, 14190-14190 (2017-02-01)
Cyclohexanone and its derivatives are very important chemicals, which are currently produced mainly by oxidation of cyclohexane or alkylcyclohexane, hydrogenation of phenols, and alkylation of cyclohexanone. Here we report that bromide salt-modified Pd/C in H
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service