Documents
BR782155
BRAND® lids for microplates BRAND®plates
plate size 96 wells, without condensation rings
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
feature
without condensation rings
manufacturer/tradename
BRAND 782155
plate size
96 wells
General description
To protect samples, reaction batches or cell cultures from both contamination and evaporation, compatible lids are available for all BRANDplates® microplates. The lids are made of crystal-clear polystyrene. They are optimally matched to the corresponding BRANDplates® microplates, so that plates with lids can also be stacked. The cut-corner sets the orientation of the lid. For 96-well standard plates, lids with condensation rings are available.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG