BR781680
BRAND® microplate BRANDplates®, pureGrade
size 384 wells, size 100 μL, flat bottom, transparent
Synonym(s):
BRANDplates®, Microplates, pureGradeS
About This Item
flat bottom
polystyrene
384 wells
Recommended Products
material
colorless wells
flat bottom
polystyrene
sterility
sterile
feature
lid
skirt (F-bottom)
packaging
pack of 50 ea
pkg of (individually packaged)
manufacturer/tradename
BRAND 781680
size
100 μL
384 wells
working volume
100 μL
color
transparent
suitability
suitable for (bacteriological assays)
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Optimal surfaces for optimal results
- Reliable quality from the cleanroom
- Tested by independent laboratories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service