Skip to Content
Merck
All Photos(1)

Key Documents

BR701353

Deep well plate 96-well

polypropylene, non-sterile, V-bottom

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.15
material:
V-bottom
polypropylene
size:
96 well
sterility:
non-sterile
Pricing and availability is not currently available.

material

V-bottom
polypropylene

sterility

non-sterile

usage

sample storage

packaging

bag of 1 ea (30 pieces)

size

96 well

well volume

2.2 mL

wells

96 wells

General description

This is a deep well plate with 96-wells and is constructed with Polypropylene polymer (PP). This is a 2.2ml, 96-well deep well plate which has a V-bottom. DNA-, DNase- and RNase-free (except 701340) 12 x 8 configuration, capacities 0.5 ml, 1.1 ml, 1.2 ml and 2.2 ml. Designed for a wide range of applications, such as High-Throughput Screening (HTS), tests requiring a mother plate, cell and tissue cultures, serial dilutions, reagent transfer and sample storage down to -80 °C (PP). Deep-well plates are manufactured according to ANSI/SLAS 1 and 4in and are stackable for easy storage.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service